Assay Details
| Gene Symbol: cu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104277.2 | NP_001097747.1 | 2 - 3 | 428 | 102 | |
| GenBank mRNA | BT023825.1 | - | 2 - 3 | 421 | 102 | |
| BT025859.1 | - | 2 - 3 | 398 | 102 | ||
More Information
Set Membership: |
|

cu
Genomic Map