Assay Details
| Gene Symbol: CG31368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169439.4 | NP_731647.3 | 6 - 7 | 3725 | 129 | |
| NM_206476.3 | NP_996198.2 | 6 - 7 | 3725 | 129 | ||
| GenBank mRNA | BT009996.1 | - | 6 - 7 | 3720 | 129 | |
More Information
Set Membership: |
|

CG31368
Genomic Map