Assay Details
| Gene Symbol: CG31368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206476.3 | NP_996198.2 | 7 - 8 | 4099 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG31368| Gene Symbol: CG31368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206476.3 | NP_996198.2 | 7 - 8 | 4099 | 86 | |
Set Membership: |
|