Assay Details
| Gene Symbol: ATP8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260138.2 | NP_001247067.1 | 18 - 19 | 4830 | 91 | |
| NM_001260139.2 | NP_001247068.1 | 17 - 18 | 4337 | 91 | ||
| NM_001260140.2 | NP_001247069.2 | 18 - 19 | 4788 | 91 | ||
| NM_001300365.1 | NP_001287294.1 | 17 - 18 | 4140 | 91 | ||
| NM_001300366.1 | NP_001287295.1 | 18 - 19 | 4575 | 91 | ||
| NM_001300367.1 | NP_001287296.1 | 18 - 19 | 4533 | 91 | ||
| NM_169450.4 | NP_731669.2 | 18 - 19 | 5027 | 91 | ||
More Information
Set Membership: |
|

ATP8B
Genomic Map