Assay Details
| Gene Symbol: CG34402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104286.2 | NP_001097756.1 | 3 - 4 | 545 | 68 | |
| NM_001104288.2 | NP_001097758.1 | 1 - 2 | 395 | 68 | ||
| NM_001300376.1 | NP_001287305.1 | 3 - 4 | 512 | 68 | ||
| GenBank mRNA | BT023111.1 | - | 1 - 2 | 395 | 68 | |
| DQ015970.1 | - | 3 - 4 | 506 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT023111.1, NM_001104288.2 transcripts.
Set Membership: |
|

CG34402
Genomic Map