Assay Details
| Gene Symbol: mRpS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275606.1 | NP_001262535.1 | 1 - 2 | 241 | 66 | |
| NM_169518.2 | NP_731803.1 | 1 - 2 | 241 | 66 | ||
| GenBank mRNA | BT001577.1 | - | 1 - 2 | 225 | 66 | |
More Information
Set Membership: |
|

mRpS21
Genomic Map