Assay Details
| Gene Symbol: rin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080168.3 | NP_524907.2 | 2 - 3 | 1779 | 68 | |
| NM_169531.2 | NP_731828.1 | 1 - 2 | 1678 | 68 | ||
| NM_169532.2 | NP_731829.1 | 3 - 4 | 2012 | 68 | ||
| NM_169533.2 | NP_731830.2 | 2 - 3 | 1779 | 68 | ||
| GenBank mRNA | AY058617.1 | - | 2 - 3 | 1766 | 68 | |
| BT011006.1 | - | 1 - 2 | 1680 | 68 | ||
More Information
Set Membership: |
|

rin
Genomic Map