Assay Details
| Gene Symbol: Lkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169541.3 | NP_731840.1 | 5 - 6 | 2128 | 100 | |
| NM_169543.3 | NP_731842.1 | 5 - 6 | 2122 | 100 | ||
More Information
Set Membership: |
|

Lkb1
Genomic Map