Assay Details
| Gene Symbol: put | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275645.1 | NP_001262574.1 | 4 - 5 | 1013 | 64 | |
| NM_001275646.1 | NP_001262575.1 | 4 - 5 | 1013 | 64 | ||
| NM_169591.3 | NP_731926.1 | 4 - 5 | 1013 | 64 | ||
| GenBank mRNA | L38495.1 | - | 4 - 5 | 986 | 64 | |
More Information
Set Membership: |
|

put
Genomic Map