Assay Details
| Gene Symbol: Meltrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032403.2 | NP_001027575.1 | 2 - 3 | 911 | 68 | |
| NM_001032404.3 | NP_001027576.2 | 2 - 3 | 911 | 68 | ||
| GenBank mRNA | AY525769.1 | - | 2 - 3 | 224 | 68 | |
More Information
Set Membership: |
|

Meltrin
Genomic Map