Assay Details
| Gene Symbol: Tm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169645.4 | NP_732013.2 | 3 - 4 | 876 | 89 | |
| GenBank mRNA | BT133082.1 | - | 2 - 3 | 835 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm2| Gene Symbol: Tm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169645.4 | NP_732013.2 | 3 - 4 | 876 | 89 | |
| GenBank mRNA | BT133082.1 | - | 2 - 3 | 835 | 89 | |
Set Membership: |
|