Assay Details
| Gene Symbol: Rbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170149.3 | NP_001163620.3 | 11 - 12 | 2630 | 52 | |
| NM_001202297.3 | NP_001189226.1 | 11 - 12 | 2660 | 52 | ||
| NM_001202298.3 | NP_001189227.1 | 11 - 12 | 2660 | 52 | ||
| NM_001275672.2 | NP_001262601.1 | 11 - 12 | 3363 | 52 | ||
| NM_001275673.2 | NP_001262602.1 | 11 - 12 | 2981 | 52 | ||
| NM_001316579.1 | NP_001303508.1 | 11 - 12 | 2552 | 52 | ||
| NM_169652.5 | NP_732028.4 | 11 - 12 | 2582 | 52 | ||
| NM_169653.4 | NP_732029.4 | 11 - 12 | 2585 | 52 | ||
| NM_169654.4 | NP_732030.4 | 11 - 12 | 3255 | 52 | ||
More Information
Set Membership: |
|

Rbp
Genomic Map