Assay Details
| Gene Symbol: Rbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202297.3 | NP_001189226.1 | 6 - 7 | 1432 | 89 | |
| NM_001202298.3 | NP_001189227.1 | 6 - 7 | 1432 | 89 | ||
| NM_001275672.2 | NP_001262601.1 | 6 - 7 | 2141 | 89 | ||
| NM_001275673.2 | NP_001262602.1 | 6 - 7 | 1786 | 89 | ||
More Information
Set Membership: |
|

Rbp
Genomic Map