Assay Details
| Gene Symbol: srp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169694.3 | NP_732098.1 | 1 - 2 | 1586 | 91 | |
| NM_169696.3 | NP_732100.2 | 1 - 2 | 1586 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srp| Gene Symbol: srp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169694.3 | NP_732098.1 | 1 - 2 | 1586 | 91 | |
| NM_169696.3 | NP_732100.2 | 1 - 2 | 1586 | 91 | ||
Set Membership: |
|