Assay Details
| Gene Symbol: srp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260199.2 | NP_001247128.1 | 5 - 6 | 1806 | 69 | |
| NM_169694.3 | NP_732098.1 | 4 - 5 | 2424 | 69 | ||
More Information
Set Membership: |
|

srp
Genomic Map