Assay Details
| Gene Symbol: srp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032019.3 | NP_001027190.1 | 7 - 8 | 2168 | 144 | |
| NM_001260199.2 | NP_001247128.1 | 7 - 8 | 2123 | 144 | ||
| NM_001275689.1 | NP_001262618.1 | 7 - 8 | 1601 | 144 | ||
| NM_001382083.1 | NP_001369017.1 | 6 - 7 | 1360 | 144 | ||
| NM_169694.3 | NP_732098.1 | 6 - 7 | 2741 | 144 | ||
| NM_169696.3 | NP_732100.2 | 6 - 7 | 2786 | 144 | ||
| GenBank mRNA | BT011543.1 | - | 7 - 8 | 2119 | 144 | |
More Information
Set Membership: |
|

srp
Genomic Map