Assay Details
| Gene Symbol: srp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032019.3 | NP_001027190.1 | 5 - 6 | 1842 | 134 | |
| NM_001275689.1 | NP_001262618.1 | 5 - 6 | 1275 | 134 | ||
| NM_001275690.1 | NP_001262619.1 | 5 - 6 | 1842 | 134 | ||
| NM_001382083.1 | NP_001369017.1 | 4 - 5 | 1034 | 134 | ||
| NM_169696.3 | NP_732100.2 | 4 - 5 | 2460 | 134 | ||
| GenBank mRNA | BT011543.1 | - | 5 - 6 | 1793 | 134 | |
More Information
Set Membership: |
|

srp
Genomic Map