Assay Details
| Gene Symbol: gish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014626.4 | NP_001014626.1 | 7 - 8 | 1872 | 128 | |
| NM_001014627.2 | NP_001014627.2 | 5 - 6 | 682 | 128 | ||
| NM_001014628.3 | NP_001014628.1 | 7 - 8 | 1227 | 128 | ||
| NM_001170157.2 | NP_001163628.1 | 6 - 7 | 1950 | 128 | ||
| NM_001260208.2 | NP_001247137.1 | 7 - 8 | 2025 | 128 | ||
| NM_001260209.2 | NP_001247138.1 | 7 - 8 | 2016 | 128 | ||
| NM_001275695.1 | NP_001262624.1 | 6 - 7 | 1668 | 128 | ||
| NM_001275696.1 | NP_001262625.1 | 6 - 7 | 1950 | 128 | ||
| NM_001382189.1 | NP_001369018.1 | 6 - 7 | 1051 | 128 | ||
| NM_001382190.1 | NP_001369019.1 | 6 - 7 | 1051 | 128 | ||
| NM_001382191.1 | NP_001369020.1 | 6 - 7 | 1051 | 128 | ||
| NM_080202.6 | NP_524941.3 | 6 - 7 | 1053 | 128 | ||
| NM_169711.5 | NP_732123.1 | 6 - 7 | 1051 | 128 | ||
| NM_169712.4 | NP_732124.3 | 6 - 7 | 1950 | 128 | ||
| NM_169713.5 | NP_732125.2 | 6 - 7 | 1668 | 128 | ||
| NM_176506.4 | NP_788683.1 | 6 - 7 | 1935 | 128 | ||
| GenBank mRNA | BT001330.1 | - | 6 - 7 | 940 | 128 | |
| BT004871.1 | - | 6 - 7 | 1910 | 128 | ||
| BT125862.1 | - | 5 - 6 | 702 | 128 | ||
More Information
Set Membership: |
|

gish
Genomic Map