Assay Details
| Gene Symbol: gish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014626.4 | NP_001014626.1 | 9 - 10 | 2313 | 61 | |
| NM_001014627.2 | NP_001014627.2 | 7 - 8 | 1123 | 61 | ||
| NM_001170157.2 | NP_001163628.1 | 8 - 9 | 2391 | 61 | ||
| NM_001260208.2 | NP_001247137.1 | 9 - 10 | 2466 | 61 | ||
| NM_001260209.2 | NP_001247138.1 | 9 - 10 | 2457 | 61 | ||
| NM_001275695.1 | NP_001262624.1 | 8 - 9 | 2109 | 61 | ||
| NM_001275696.1 | NP_001262625.1 | 8 - 9 | 2391 | 61 | ||
| NM_001382190.1 | NP_001369019.1 | 8 - 9 | 1492 | 61 | ||
| NM_080202.6 | NP_524941.3 | 8 - 9 | 1494 | 61 | ||
| NM_169711.5 | NP_732123.1 | 8 - 9 | 1492 | 61 | ||
| NM_169712.4 | NP_732124.3 | 8 - 9 | 2391 | 61 | ||
| NM_169713.5 | NP_732125.2 | 8 - 9 | 2109 | 61 | ||
| NM_176506.4 | NP_788683.1 | 8 - 9 | 2376 | 61 | ||
| GenBank mRNA | BT004871.1 | - | 8 - 9 | 2351 | 61 | |
| BT125862.1 | - | 7 - 8 | 1143 | 61 | ||
More Information
Set Membership: |
|

gish
Genomic Map