Assay Details
| Gene Symbol: gish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014626.4 | NP_001014626.1 | 10 - 11 | 2392 | 88 | |
| NM_001014627.2 | NP_001014627.2 | 8 - 9 | 1202 | 88 | ||
| NM_001170157.2 | NP_001163628.1 | 9 - 10 | 2470 | 88 | ||
| NM_001260209.2 | NP_001247138.1 | 10 - 11 | 2536 | 88 | ||
| NM_001275695.1 | NP_001262624.1 | 9 - 10 | 2188 | 88 | ||
| NM_001275696.1 | NP_001262625.1 | 9 - 10 | 2470 | 88 | ||
| NM_001382189.1 | NP_001369018.1 | 10 - 11 | 1649 | 88 | ||
| NM_001382190.1 | NP_001369019.1 | 9 - 10 | 1571 | 88 | ||
| NM_080202.6 | NP_524941.3 | 9 - 10 | 1573 | 88 | ||
| NM_169711.5 | NP_732123.1 | 9 - 10 | 1571 | 88 | ||
| NM_169712.4 | NP_732124.3 | 9 - 10 | 2470 | 88 | ||
| NM_169713.5 | NP_732125.2 | 9 - 10 | 2188 | 88 | ||
| NM_176506.4 | NP_788683.1 | 9 - 10 | 2455 | 88 | ||
| GenBank mRNA | BT004871.1 | - | 9 - 10 | 2430 | 88 | |
| BT125862.1 | - | 8 - 9 | 1222 | 88 | ||
More Information
Set Membership: |
|

gish
Genomic Map