Assay Details
| Gene Symbol: gish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014626.4 | NP_001014626.1 | 5 - 6 | 1597 | 88 | |
| NM_001014627.2 | NP_001014627.2 | 3 - 4 | 407 | 88 | ||
| NM_001014628.3 | NP_001014628.1 | 5 - 6 | 952 | 88 | ||
| NM_001170157.2 | NP_001163628.1 | 4 - 5 | 1675 | 88 | ||
| NM_001260208.2 | NP_001247137.1 | 5 - 6 | 1750 | 88 | ||
| NM_001260209.2 | NP_001247138.1 | 5 - 6 | 1741 | 88 | ||
| NM_001275695.1 | NP_001262624.1 | 4 - 5 | 1393 | 88 | ||
| NM_001275696.1 | NP_001262625.1 | 4 - 5 | 1675 | 88 | ||
| NM_080202.6 | NP_524941.3 | 4 - 5 | 778 | 88 | ||
| NM_169712.4 | NP_732124.3 | 4 - 5 | 1675 | 88 | ||
| NM_169713.5 | NP_732125.2 | 4 - 5 | 1393 | 88 | ||
| GenBank mRNA | BT001330.1 | - | 4 - 5 | 665 | 88 | |
| BT004871.1 | - | 4 - 5 | 1635 | 88 | ||
More Information
Set Membership: |
|

gish
Genomic Map