Assay Details
| Gene Symbol: Dscam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104355.2 | NP_001097825.1 | 11 - 12 | 2047 | 71 | |
| NM_001104356.2 | NP_001097826.1 | 10 - 11 | 1642 | 71 | ||
| NM_169761.2 | NP_732242.2 | 10 - 11 | 1642 | 71 | ||
| NM_206504.2 | NP_996226.2 | 10 - 11 | 1642 | 71 | ||
| GenBank mRNA | BT014656.1 | - | 10 - 11 | 1590 | 71 | |
More Information
Set Membership: |
|

Dscam3
Genomic Map