Assay Details
| Gene Symbol: Dscam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104355.2 | NP_001097825.1 | 28 - 29 | 5670 | 85 | |
| NM_169761.2 | NP_732242.2 | 28 - 29 | 5193 | 85 | ||
| NM_206504.2 | NP_996226.2 | 28 - 29 | 5193 | 85 | ||
More Information
Set Membership: |
|

Dscam3
Genomic Map