Assay Details
| Gene Symbol: Dscam3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104355.2 | NP_001097825.1 | 6 - 7 | 1267 | 104 | |
| NM_001104356.2 | NP_001097826.1 | 5 - 6 | 862 | 104 | ||
| NM_169761.2 | NP_732242.2 | 5 - 6 | 862 | 104 | ||
| NM_206504.2 | NP_996226.2 | 5 - 6 | 862 | 104 | ||
| GenBank mRNA | BT014656.1 | - | 5 - 6 | 810 | 104 | |
More Information
Set Membership: |
|

Dscam3
Genomic Map