Assay Details
| Gene Symbol: 14-3-3epsilon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169796.2 | NP_732309.1 | 2 - 3 | 691 | 71 | |
| NM_169797.3 | NP_732310.1 | 2 - 3 | 847 | 71 | ||
| NM_169798.3 | NP_732311.1 | 2 - 3 | 847 | 71 | ||
| NM_169799.2 | NP_732312.1 | 2 - 3 | 691 | 71 | ||
| GenBank mRNA | BT099556.1 | - | 2 - 3 | 638 | 71 | |
| U84897.1 | - | 2 - 3 | 624 | 71 | ||
More Information
Set Membership: |
|

14-3-3epsilon
Genomic Map