Assay Details
| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170177.2 | NP_001163648.1 | 8 - 9 | 2346 | 84 | |
| NM_169819.2 | NP_732347.1 | 7 - 8 | 2331 | 84 | ||
| NM_169820.2 | NP_732348.1 | 7 - 8 | 3932 | 84 | ||
| NM_206512.2 | NP_996234.1 | 6 - 7 | 1637 | 84 | ||
| NM_206513.2 | NP_996235.1 | 6 - 7 | 2194 | 84 | ||
| NM_206515.2 | NP_996237.1 | 6 - 7 | 1786 | 84 | ||
| NM_206516.2 | NP_996238.1 | 6 - 7 | 1634 | 84 | ||
| GenBank mRNA | BT024450.1 | - | 8 - 9 | 2346 | 84 | |
More Information
Set Membership: |
|

fru
Genomic Map