Assay Details
| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169822.2 | NP_732350.1 | 5 - 6 | 1452 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fru| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169822.2 | NP_732350.1 | 5 - 6 | 1452 | 87 | |
Set Membership: |
|