Assay Details
| Gene Symbol: cry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169852.2 | NP_732407.1 | 1 - 2 | 326 | 70 | |
| GenBank mRNA | AB018400.1 | - | 1 - 2 | 139 | 70 | |
| AF099734.1 | - | 1 - 2 | 178 | 70 | ||
| AY051514.1 | - | 1 - 2 | 254 | 70 | ||
More Information
Set Membership: |
|

cry
Genomic Map