Assay Details
| Gene Symbol: cry | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169852.2 | NP_732407.1 | 3 - 4 | 1592 | 93 | |
| GenBank mRNA | AB018400.1 | - | 3 - 4 | 1405 | 93 | |
| AF099734.1 | - | 3 - 4 | 1444 | 93 | ||
| AY051514.1 | - | 3 - 4 | 1520 | 93 | ||
More Information
Set Membership: |
|

cry
Genomic Map