Assay Details
| Gene Symbol: Stat92E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275830.1 | NP_001262759.1 | 2 - 3 | 647 | 58 | |
| NM_001275831.1 | NP_001262760.1 | 2 - 3 | 803 | 58 | ||
| NM_001275833.1 | NP_001262762.1 | 2 - 3 | 803 | 58 | ||
| NM_169899.3 | NP_732516.2 | 2 - 3 | 803 | 58 | ||
| NM_206520.2 | NP_996242.1 | 2 - 3 | 647 | 58 | ||
| NM_206521.2 | NP_996243.1 | 2 - 3 | 647 | 58 | ||
| GenBank mRNA | BT021314.1 | - | 2 - 3 | 648 | 58 | |
| U40070.1 | - | 2 - 3 | 622 | 58 | ||
More Information
Set Membership: |
|

Stat92E
Genomic Map