Assay Details
| Gene Symbol: Stat92E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014643.2 | NP_001014643.1 | 2 - 3 | 1415 | 69 | |
| NM_001275828.1 | NP_001262757.1 | 2 - 3 | 1259 | 69 | ||
| NM_001275829.1 | NP_001262758.1 | 2 - 3 | 1415 | 69 | ||
| NM_001275830.1 | NP_001262759.1 | 3 - 4 | 1672 | 69 | ||
| NM_001275831.1 | NP_001262760.1 | 3 - 4 | 1828 | 69 | ||
| NM_001275832.1 | NP_001262761.1 | 2 - 3 | 1259 | 69 | ||
| NM_001275833.1 | NP_001262762.1 | 3 - 4 | 1828 | 69 | ||
| NM_169899.3 | NP_732516.2 | 3 - 4 | 1828 | 69 | ||
| NM_169900.3 | NP_732517.2 | 2 - 3 | 1415 | 69 | ||
| NM_206520.2 | NP_996242.1 | 3 - 4 | 1672 | 69 | ||
| NM_206521.2 | NP_996243.1 | 3 - 4 | 1672 | 69 | ||
| GenBank mRNA | BT021314.1 | - | 3 - 4 | 1673 | 69 | |
| U40070.1 | - | 3 - 4 | 1647 | 69 | ||
More Information
Set Membership: |
|

Stat92E
Genomic Map