Assay Details
| Gene Symbol: Atpalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170196.3 | NP_001163667.1 | 5 - 6 | 2330 | 64 | |
| NM_001275861.1 | NP_001262790.1 | 5 - 6 | 2396 | 64 | ||
| NM_001275862.2 | NP_001262791.1 | 6 - 7 | 2474 | 64 | ||
| NM_169936.3 | NP_732572.1 | 5 - 6 | 2311 | 64 | ||
| NM_169937.3 | NP_732573.1 | 4 - 5 | 2614 | 64 | ||
| NM_169938.3 | NP_732574.1 | 5 - 6 | 2330 | 64 | ||
| NM_169939.2 | NP_732575.1 | 5 - 6 | 2714 | 64 | ||
| NM_206525.3 | NP_996247.1 | 4 - 5 | 2614 | 64 | ||
| NM_206526.3 | NP_996248.1 | 5 - 6 | 2330 | 64 | ||
| NM_206527.3 | NP_996249.1 | 5 - 6 | 2330 | 64 | ||
| NM_206528.3 | NP_996250.1 | 6 - 7 | 2839 | 64 | ||
| GenBank mRNA | BT044212.1 | - | 5 - 6 | 2687 | 64 | |
| BT133381.1 | - | 5 - 6 | 2298 | 64 | ||
More Information
Set Membership: |
|

Atpalpha
Genomic Map