Assay Details
| Gene Symbol: Atpalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170196.3 | NP_001163667.1 | 8 - 9 | 2818 | 86 | |
| NM_001275861.1 | NP_001262790.1 | 8 - 9 | 2884 | 86 | ||
| NM_001275862.2 | NP_001262791.1 | 9 - 10 | 2962 | 86 | ||
| NM_169936.3 | NP_732572.1 | 8 - 9 | 2799 | 86 | ||
| NM_169937.3 | NP_732573.1 | 7 - 8 | 3102 | 86 | ||
| NM_169938.3 | NP_732574.1 | 8 - 9 | 2818 | 86 | ||
| NM_169939.2 | NP_732575.1 | 7 - 8 | 3393 | 86 | ||
| NM_206525.3 | NP_996247.1 | 7 - 8 | 3102 | 86 | ||
| NM_206526.3 | NP_996248.1 | 8 - 9 | 2818 | 86 | ||
| NM_206527.3 | NP_996249.1 | 8 - 9 | 2818 | 86 | ||
| NM_206528.3 | NP_996250.1 | 9 - 10 | 3327 | 86 | ||
| GenBank mRNA | BT044212.1 | - | 7 - 8 | 3366 | 86 | |
| BT133381.1 | - | 8 - 9 | 2786 | 86 | ||
More Information
Set Membership: |
|

Atpalpha
Genomic Map