Assay Details
| Gene Symbol: SNF4Agamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080509.3 | NP_536757.2 | 3 - 4 | 744 | 88 | |
| NM_169946.2 | NP_732594.1 | 3 - 4 | 915 | 88 | ||
| GenBank mRNA | BT003626.1 | - | 744 | 88 | ||
| BT010233.1 | - | 3 - 4 | 913 | 88 | ||
| BT120301.1 | - | 3 - 4 | 763 | 88 | ||
More Information
Set Membership: |
|

SNF4Agamma
Genomic Map