Assay Details
| Gene Symbol: bap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169958.2 | NP_732637.1 | 1 - 2 | 579 | 106 | |
| GenBank mRNA | BT001561.1 | - | 1 - 2 | 572 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

bap| Gene Symbol: bap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169958.2 | NP_732637.1 | 1 - 2 | 579 | 106 | |
| GenBank mRNA | BT001561.1 | - | 1 - 2 | 572 | 106 | |
Set Membership: |
|