Assay Details
| Gene Symbol: CG31343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169964.3 | NP_732652.1 | 4 - 5 | 2691 | 65 | |
| GenBank mRNA | AY095013.1 | - | 4 - 5 | 2667 | 65 | |
| Gene Symbol: Hsp70Bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT011379.1 | - | 5211 | 65 | ||
More Information
Set Membership: |
|

CG31343
Genomic Map