Assay Details
| Gene Symbol: lqfR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170009.4 | NP_732734.3 | 3 - 4 | 909 | 160 | |
| NM_170011.3 | NP_732736.2 | 3 - 4 | 909 | 160 | ||
| GenBank mRNA | AY060606.1 | - | 3 - 4 | 877 | 160 | |
More Information
Set Membership: |
|

lqfR
Genomic Map