Assay Details
| Gene Symbol: orb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170228.1 | NP_001163699.1 | 2 - 3 | 370 | 59 | |
| NM_170062.2 | NP_732848.1 | 1 - 2 | 535 | 59 | ||
| NM_170063.2 | NP_732849.2 | 1 - 2 | 535 | 59 | ||
| GenBank mRNA | BT001315.1 | - | 2 - 3 | 371 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT001315.1, NM_001170228.1 transcripts.
Set Membership: |
|

orb
Genomic Map