Assay Details
Gene Symbol: Irk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170229.2 | NP_001163700.1 | 1 - 2 | 250 | 60 | |
NM_170076.2 | NP_732871.1 | 1 - 2 | 250 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
