Assay Details
| Gene Symbol: jar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014647.3 | NP_001014647.2 | 14 - 15 | 3600 | 69 | |
| NM_001014648.3 | NP_001014648.2 | 14 - 15 | 3600 | 69 | ||
| NM_001014650.3 | NP_001014650.2 | 15 - 16 | 3645 | 69 | ||
| NM_001275976.1 | NP_001262905.1 | 15 - 16 | 3403 | 69 | ||
| NM_001275977.2 | NP_001262906.1 | 14 - 15 | 4067 | 69 | ||
| NM_001300572.1 | NP_001287501.1 | 13 - 14 | 3555 | 69 | ||
| NM_079754.4 | NP_524478.4 | 14 - 15 | 3600 | 69 | ||
| NM_170138.3 | NP_732976.1 | 14 - 15 | 3358 | 69 | ||
| GenBank mRNA | BT133083.1 | - | 14 - 15 | 3597 | 69 | |
More Information
Set Membership: |
|

jar
Genomic Map