Assay Details
| Gene Symbol: puf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275993.2 | NP_001262922.1 | 16 - 17 | 9430 | 104 | |
| NM_001275994.2 | NP_001262923.1 | 15 - 16 | 9509 | 104 | ||
| NM_143018.4 | NP_651275.1 | 15 - 16 | 9509 | 104 | ||
More Information
Set Membership: |
|

puf
Genomic Map