Assay Details
| Gene Symbol: puf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275993.2 | NP_001262922.1 | 19 - 20 | 10936 | 107 | |
| NM_001275994.2 | NP_001262923.1 | 18 - 19 | 11015 | 107 | ||
| NM_001300579.1 | NP_001287508.1 | 18 - 19 | 11015 | 107 | ||
| NM_143018.4 | NP_651275.1 | 18 - 19 | 11015 | 107 | ||
| NM_170158.3 | NP_733022.2 | 18 - 19 | 11015 | 107 | ||
More Information
Set Membership: |
|

puf
Genomic Map