Assay Details
| Gene Symbol: ash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260363.1 | NP_001247292.1 | 3 - 4 | 735 | 93 | |
| NM_170160.2 | NP_733024.1 | 3 - 4 | 877 | 93 | ||
| NM_176558.3 | NP_788735.1 | 5 - 6 | 1413 | 93 | ||
| GenBank mRNA | AY051803.1 | - | 5 - 6 | 1366 | 93 | |
| BT011328.1 | - | 3 - 4 | 767 | 93 | ||
| U73809.2 | - | 5 - 6 | 1366 | 93 | ||
More Information
Set Membership: |
|

ash2
Genomic Map