Assay Details
| Gene Symbol: slo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014654.3 | NP_001014654.1 | 16 - 17 | 2767 | 84 | |
| NM_001014659.3 | NP_001014659.1 | 17 - 18 | 2806 | 84 | ||
| NM_001014661.4 | NP_001014661.2 | 17 - 18 | 2523 | 84 | ||
| NM_001170241.3 | NP_001163712.1 | 18 - 19 | 3051 | 84 | ||
| NM_001275995.1 | NP_001262924.1 | 18 - 19 | 2733 | 84 | ||
| NM_170164.4 | NP_733029.1 | 17 - 18 | 2508 | 84 | ||
| NM_206566.4 | NP_996289.1 | 17 - 18 | 2508 | 84 | ||
More Information
Set Membership: |
|

slo
Genomic Map