Assay Details
| Gene Symbol: slo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014653.3 | NP_001014653.1 | 7 - 8 | 1781 | 79 | |
| NM_001014654.3 | NP_001014654.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014655.3 | NP_001014655.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014656.3 | NP_001014656.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014657.3 | NP_001014657.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014658.3 | NP_001014658.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014659.3 | NP_001014659.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014660.3 | NP_001014660.1 | 7 - 8 | 1781 | 79 | ||
| NM_001014662.3 | NP_001014662.1 | 7 - 8 | 1781 | 79 | ||
| NM_001275996.1 | NP_001262925.1 | 7 - 8 | 1781 | 79 | ||
| NM_001275997.1 | NP_001262926.1 | 8 - 9 | 1522 | 79 | ||
| NM_001275998.1 | NP_001262927.1 | 8 - 9 | 1522 | 79 | ||
| NM_001275999.1 | NP_001262928.1 | 8 - 9 | 1522 | 79 | ||
| NM_079762.4 | NP_524486.2 | 7 - 8 | 1781 | 79 | ||
| NM_170164.4 | NP_733029.1 | 8 - 9 | 1522 | 79 | ||
| NM_206566.4 | NP_996289.1 | 8 - 9 | 1522 | 79 | ||
More Information
Set Membership: |
|

slo
Genomic Map