Assay Details
| Gene Symbol: PIG-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170181.3 | NP_733060.1 | 1 - 2 | 160 | 76 | |
| GenBank mRNA | AY061834.1 | - | 1 - 2 | 132 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIG-S| Gene Symbol: PIG-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170181.3 | NP_733060.1 | 1 - 2 | 160 | 76 | |
| GenBank mRNA | AY061834.1 | - | 1 - 2 | 132 | 76 | |
Set Membership: |
|