Assay Details
| Gene Symbol: PIG-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170181.3 | NP_733060.1 | 2 - 3 | 252 | 100 | |
| GenBank mRNA | AY061834.1 | - | 2 - 3 | 224 | 100 | |
More Information
Set Membership: |
|

PIG-S
Genomic Map