Assay Details
| Gene Symbol: Tl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276066.2 | NP_001262995.1 | 3 - 4 | 1802 | 67 | |
| NM_079794.3 | NP_524518.1 | 3 - 4 | 1802 | 67 | ||
| NM_170287.3 | NP_733166.1 | 4 - 5 | 1871 | 67 | ||
| GenBank mRNA | BT150335.1 | - | 3 - 4 | 1797 | 67 | |
| M19969.1 | - | 3 - 4 | 1797 | 67 | ||
More Information
Set Membership: |
|

Tl
Genomic Map