Assay Details
| Gene Symbol: Rb97D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014671.2 | NP_001014671.1 | 3 - 4 | 670 | 64 | |
| NM_001014672.2 | NP_001014672.1 | 3 - 4 | 670 | 64 | ||
| NM_001014673.2 | NP_001014673.1 | 3 - 4 | 670 | 64 | ||
| NM_001014674.2 | NP_001014674.1 | 3 - 4 | 670 | 64 | ||
| NM_001014675.2 | NP_001014675.1 | 3 - 4 | 670 | 64 | ||
| NM_001014676.2 | NP_001014676.1 | 3 - 4 | 670 | 64 | ||
| NM_001014677.2 | NP_001014677.1 | 3 - 4 | 670 | 64 | ||
| NM_079796.5 | NP_524520.1 | 3 - 4 | 670 | 64 | ||
| NM_170293.3 | NP_733172.1 | 3 - 4 | 670 | 64 | ||
| GenBank mRNA | BT001320.1 | - | 3 - 4 | 670 | 64 | |
More Information
Set Membership: |
|

Rb97D
Genomic Map