Assay Details
| Gene Symbol: TfIIA-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079743.4 | NP_524467.1 | 1 - 2 | 179 | 73 | |
| GenBank mRNA | BT044069.1 | - | 1 - 2 | 154 | 73 | |
| KX531771.1 | - | 1 - 2 | 74 | 73 | ||
More Information
Set Membership: |
|

TfIIA-S
Genomic Map